Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant Astrocytic Phosphoprotein PEA-15 (from E. coli)
Human recombinant Astrocytic Phosphoprotein PEA-15 (from E. coli)
Catalog # PRSI92-118
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Astrocytic Phosphoprotein PEA-15 (from E. coli)
Catalog # PRSI92-118
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    15 kDa Phosphoprotein Enriched in Astrocytes|PEA15|Phosphoprotein Enriched in Diabetes|Astrocytic Phosphoprotein PEA-15|PED
  • Protein/peptide name:
    Astrocytic Phosphoprotein PEA-15
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    15.3 kD
  • Sequence:
    Met1-Ala130
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Astrocyticphosphoprotein PEA-15 (PEA15) is a death effector domain (DED)-containing protein. PEA15 is mainly expressed in the central nervous system, principally in astrocytes. Increased PEA15 levels affect tumourigenesis and cancer progression. PEA15 is overexpressed in breast cancers and gliomas as well as in type 2 diabetes. PEA15 blocks Ras-mediated inhibition of integrin activation and modulates the ERK MAP kinase cascade. PEA15 also inhibits RPS6KA3 activities by holding it in the cytoplasm. In addition, PEA15 inhibits both TNFRSF6 and TNFRSF1A mediated CASP8 activity and apoptosis. At present, PEA15 expression is also a significant prognostic marker in ovarian cancer.

  • This recombinant protein can be used for biological assays. For research use only.

This recombinant protein can be used for biological assays. For research use only.