Human recombinant BDH1 (from E. coli)
Catalog # PRSI92-066
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:BDH|D-Beta-Hydroxybutyrate Dehydrogenase Mitochondrial|3-Hydroxybutyrate Dehydrogenase
- Protein/peptide name:BDH1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:40.3 kD
- Sequence:Met1-Arg343
- Formulation:Supplied as a 0.2 um filtered solution of 50mM Tris, 1mM DTT, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:used for biological assays
Specifications
About this item
D-Beta-Hydroxybutyrate Dehydrogenase Mitochondrial (BDH) is a member of the short-chain dehydrogenases/reductases (SDR) family. BDH is localized in the mitochondrion matrix. BDH forms a homotetrameric lipid-requiring enzyme of the mitochondrial membrane and has a specific necessity for phosphatidylcholine for optimal enzymatic activity. BDH catalyzes the interconversion of acetoacetate and (R)-3-hydroxybutyrate, the 2 main ketone bodies formed during fatty acid catabolism.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag