Human recombinant PADI4 (from E. coli)
Catalog # PRSI92-065
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:PDI5|HL-60 PAD|PADI5|Protein-arginine Deiminase type IV|PADI4|PDI4|Peptidylarginine Deiminase IV|Protein-Arginine Deiminase Type-4
- Protein/peptide name:PADI4
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:76.2 kD
- Sequence:Met1-Pro663
- Formulation:Supplied as a 0.2 um filtered solution of 50mM Tris, 500mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:used for biological assays
Specifications
About this item
Protein-Arginine Deiminase Type-4 (PADI4) belongs to the Protein Arginine Deiminase family. PADI4 is expressed in eosinophils and neutrophils, but it is not expressed in peripheral monocytes or lymphocytes. PADI4 catalyzes the citrullination/deimination of arginine residues of proteins. In addition, PADI4 may play an important role in tumourigenesis. Genetic variations in PADI4 are a cause of susceptibility to rheumatoid arthritis (RA), which is a systemic inflammatory disease with autoimmune features and a complex genetic component.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag