Order Entry
Netherlands
ContactUsLinkComponent
 
Human Recombinant 14-3-3 epsilon (from E. coli)
  PRSI91-975
 :  
Human Recombinant 14-3-3 epsilon (from E. coli)
  PRSI91-975
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-GST tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    14-3-3 Protein Epsilon|14-3-3E|YWHAE
  • Protein/peptide name:
    14-3-3 epsilon
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    55.98 kD
  • Sequence:
    Met1-Gln255
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

14-3-3 Protein Epsilon acts as an adapter protein implicated in the regulation of a large spectrum of both general and specialized signaling pathways. 14-3-3 protein epsilon binds to a large number of partners, usually by recognition of a phophoserine or phosphothreonine motif. Binding generally results in the modulation of the activity of the binding partner.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-GST tag