Human recombinant IL-10 Receptor beta (from Cells)
Catalog # PRSI91-818
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Interleukin-10 receptor subunit 2|Interleukin-10 receptor subunit beta(IL10RB)|Cytokine receptor family 2 member 4|IL-10RB|Cytokine receptor class-II member 4
- Protein/peptide name:IL-10 Receptor beta
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:24.5 kD
- Sequence:Met20-Ser220
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
IL10RB is a single- pass type I membrane protein and contains two fibronectin type-III domains. It is an accessory chain which is essential for the active interleukin 10 receptor complex. Coexpression of IL10RB and IL10RA proteins has been shown to be required for IL10-induced signal transduction. Defects in IL10RB are the cause of inflammatory bowel disease type 25 (IBD25) which is a chronic, relapsing inflammation of the gastrointestinal tract with a complex etiology.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag