Human recombinant TINAGL1 (from cells)
PRSI91-791
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Oxidized LDL-responsive gene 2 protein|TIN Ag-related protein|OLRG2 and TINAGL. belongs to the peptidase C1 family.|LCN7|GIS5|Tubulointerstitial nephritis antigen-related protein|also known as Glucocorticoid-inducible protein 5|Tubulointerstitial nephritis antigen-like (TINAGL1)
- Protein/peptide name:TINAGL1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:51.1 kD
- Sequence:Ala22-His467
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,1mM EDTA,2m MDTT,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
TINAGL1 is a secreted protein and contains one SMB (somatomedin-B) domain. TINAGL1 is expressed in aorta, lymph nodes, heart, bone marrow, placenta, lung, kidney, skeletal muscle, pancreas, thyroid, adrenal gland and so on. TINAGL1 may be implicated in the adrenocortical zonation and in mechanisms for repressing the CYP11B1 gene expression in adrenocortical cells.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag