Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant Cadherin-11 (from Cells)
Human recombinant Cadherin-11 (from Cells)
Catalog # PRSI91-753
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Cadherin-11 (from Cells)
Catalog # PRSI91-753
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    OB-Cadherin|Cadherin 11 Type 2 OB-cadherin (Osteoblast)|CDH11|Cadherin 11 Type 2 OB-Cadherin (Osteoblast) Isoform CRA_c
  • Protein/peptide name:
    Cadherin-11
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    66.63 kD
  • Sequence:
    Phe23-Thr617
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Cadherin-11 is a type II classical cadherin member of the cadherin superfamily of integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Cadherins interact with themselves in a homophilic manner in connecting cells, and thus contribute to the sorting of heterogeneous cell types. Cadherin-11 contains five cadherin domains and is mainly expressed in the brain. Mature cadherin proteins consists of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small highly conserved C-terminal cytoplasmic domain. It is shown that Cadherin-11 is a viable molecular target for therapeutic intervention in Glioblastoma multiforme.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag