Human recombinant reticulocalbin 3 (from cells)
Catalog # PRSI91-752
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:RCN3|EF-Hand Calcium-Binding Protein RLP49|Reticulocalbin-3
- Protein/peptide name:Reticulocalbin 3
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:36.2 kD
- Sequence:Lys21-Leu328
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl,150mM NaCl,1mM DTT,10%Glycerol,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Reticulocalbin-3 is a member of the CREC family that is involved in the secretory pathway. Members of the CREC family consist of a number of multiple EF-hand domains. Reticulocalbin-3 localizes to the endoplasmic reticulum lumen. The bioinformaticanalysis of Reticulocalbin-3 reveal that it is a putative Ca2+-binding protein. In addition, it has been shown that autoactivation and secretion of PACE4 was increased upon co-expression with Reticulocalbin-3. The proPACE4-RCN-3 complex is plays an important role in the biosynthesis of PACE4.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag