Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant IFN alpha 6 (from cells)
Human recombinant IFN alpha 6 (from cells)
Catalog # PRSI91-559
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IFN alpha 6 (from cells)
Catalog # PRSI91-559
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Interferon Alpha-K|IFN-Alpha-6|IFNA6|Interferon Alpha-6|LeIF K|Interferon Alpha-54
  • Protein/peptide name:
    IFN alpha 6
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    21.1 kD
  • Sequence:
    Ser21-Glu189
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Interferon alpha-6 (IFN- alpha6) is a secreted protein which belongs to the alpha/ beta interferon family. IFN- alpha6 is produced by macrophages, expressed at low level, only 1.0% of the average gene in this release. IFN- alpha6 contains interferon alpha, beta and delta domain. IFN- alpha has antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag