Human recombinant NGR (from cells)
PRSI91-545
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:NgR|NOGOR|Nogo Receptor|RTN4R|Nogo-66 Receptor|Reticulon-4 Receptor
- Protein/peptide name:NGR
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:46.32 kD
- Sequence:Cys27-Ser447
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Nogo Receptor (NgR) is a glycosylphosphoinositol (GPI)-anchored protein that belongs to the Nogo recptor family. Human NgR is predominantly expressed in neurons and their axons in the central nervous systems. As a receptor for myelin-derived proteins Nogo, myelin-associated glycoprotein (MAG) and myelin oligodendrocyte glycoprotein (OMG), NgR mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult central nervous system. NgR may be proposed as a potential drug target for treatment of various neurological conditions. Additionally, NgR may play a role in regulating the function of gap junctions.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag