Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant CEACAM8 (from cells)
Human recombinant CEACAM8 (from cells)
  PRSI91-520
 :  
Human recombinant CEACAM8 (from cells)
  PRSI91-520
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Carcinoembryonic Antigen-Related Cell Adhesion Molecule 8|Carcinoembryonic Antigen CGM6|Non-Specific Cross-Reacting Antigen NCA-95|CEACAM8|CD66b|CGM6|CD67 Antigen
  • Protein/peptide name:
    CEACAM8
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    13.03 kD
  • Sequence:
    Gln35-His141
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

 

 

Carcinoembryonic Antigen-Related Cell Adhesion Molecule 8 (CEACAM8) is a single chain, GPI-anchored, highly glycosylated protein which belongs to the immunoglobulin superfamily and the carcinoembryonic antigen(CEA) family. CEACAM8 is expressed by neutrophils and eosinophils, and serves as a binding partner for CEACAM-6 and Galectin-3. It contains two Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. Mature human CEACAM8 is a 287 amino acid GPI-linked glycoprotein. CEACAM family members are a set of widely expressed proteins involved in several biological functions, including cell adhesion, migration, signal transduction, and the regulation of gene expression. Abnormal overexpression and downregulation of some CEACAMs have been described in tumor cells.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag