Human recombinant Annexin A2 (from E. coli)
Catalog # PRSI91-185
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:PAP-IV|p36|Lipocortin II|Protein I|LPC2D|Placental anticoagulant protein IV|Calpactin I heavy chain|CAL1H|ANX2L4|Calpactin-1 heavy chain|Annexin II|ANX2|Chromobindin-8|Annexin-2
- Protein/peptide name:Annexin A2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:38.6 kD
- Sequence:Ser2-Asp339
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, 1mM EDTA, pH 7.5 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Annexin A2 (ANXA2) is a member of the annexin family and has roles in the regulation of cellular growth and in signal transduction pathways. ANXA2 protein is associated with sickle cell osteonecrosis and the expression reduce of ANXA2 is associated with osteosarcoma metastases. ANXA2 functions as an autocrine factor, it can increases osteoclast formation and bone resorption. ANXA2 is involved in muscular dystrophies. In humans, the up-regulation of ANXA2 is related with colon adenocarcinoma cell differentiation.
- This recombinant protein can be used for biological assays. For research use only.