Human recombinant ARC (from E. coli)
Catalog # PRSI91-177
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-GST tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Muscle-Enriched Cytoplasmic Protein|NOP|Nucleolar Protein of 30 kDa|Nop30|Myp|NOL3|Apoptosis Repressor With CARD|ARC|Nucleolar Protein 3
- Protein/peptide name:ARC
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:48.92 kD
- Sequence:Met1-Ser208
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl,350mM NaCl,1mM DTT,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Nucleolar Protein 3 is encoded by NOL3 gene; multiple transcript variants encoding different isoforms have been found for this gene. So far, Nucleolar protein 3 has show to have two Isoforms. Isoform 1 may be involved in RNA splicing.Isoform 2 may inhibit apoptosis.It has been shown to down-regulate the enzyme activities of caspase 2, caspase 8 and tumor protein p53.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag