Order Entry
Netherlands
ContactUsLinkComponent
 
Human recombinant CXCL12 (from E. coli)
  PRSI91-104
 :  
Human recombinant CXCL12 (from E. coli)
  PRSI91-104
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    SDF1B|Stromal Cell-Derived Factor 1|SDF1|CXCL12|Pre-B Cell Growth-Stimulating Factor|hSDF-1|IRH|SDF1A|SDF-1(22-89)|PBSF|SDF-1|hIRH|C-X-C Motif Chemokine 12|Intercrine Reduced in Hepatomas
  • Protein/peptide name:
    CXCL12
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    8.12 kD
  • Sequence:
    Lys22-Lys89
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

 

 

Stromal Cell-Derived Factor-1 (SDF-1) is a chemokine member of the intercrine family. SDF1 is expressed as five isoforms that differ only in the C terminal tail. SDF1 alpha and SDF1 beta are identical except for the four residues present in the C-terminus of SDF1 beta but absent from SDF1 alpha. SDF1 isoforms interact with CXCR4 and CXCR7 receptors on the cell surface, and can also bind syndecan4. SDF1 is known to influence lymphopoiesis, regulate patterning and cell number of neural progenitors, and promote angiogenesis. It also enhances the survival of myeloid progenitor cells.

  • This recombinant protein can be used for biological assays. For research use only.