Human recombinant BCL-xL (from E. coli)
PRSI91-087
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:BCLX|Apoptosis Regulator Bcl-X|BCL2L1|BCL2L|Bcl2-L-1|Bcl-2-like Protein 1
- Protein/peptide name:BCL-xL
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:24.85 kD
- Sequence:Met1-Arg212
- Formulation:Supplied as a 0.2 um filtered solution of 20mM HEPES, 50mM KCl, 20% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Bcl-2-Like Protein 1 (BCL2L1) is a member of the Bcl-2 family. BCL2L1 is expressed at high levels in cells that undergo a high rate of turnover, such as developing lymphocytes. BCL2L1 is a mitochondrial membrane protein. BCL2L1 contains four motifs, BH1, BH2 and BH4. The BH4 motif is required for anti-apoptotic activity. The BH1 and BH2 motifs are required for both heterodimerization with other Bcl-2 family members and for repression of cell death. BCL2L1 regulates cell death by blocking the voltage-dependent anion channnel (VDAC) and preventing the release of the caspase activator, CYC1, from the mitochondrial membrane. In addition, BCL2L1 promotes apoptosis.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag