Human Recombinant CCL27 (from E. coli)
Catalog # PRSI91-052
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:ILC|CC Chemokine ILC|CTACK|SCYA27|Cutaneous T-Cell-Attracting Chemokine|IL-11 R-Alpha-Locus Chemokine|Small-Inducible Cytokine A27|Eskine|C-C Motif Chemokine 27|Skinkine|CCL27
- Protein/peptide name:CCL27
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:10.1 kD
- Sequence:Phe25-Gly112
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 500mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Human Chemokine (C-C Motif) Ligand 27 (CCL27) is a small cytokine that is a member of the CC chemokine family; it is expressed in numerous tissues, including gonads, thymus, placenta and skin. CCL27 elicits its chemotactic effects by binding to the chemokine receptor CCR10. Predominantly expressed in the skin, CCL27 is associated with T cell-mediated inflammation of the skin. Human and Mouse CCL27 share 84% sequence identity in the mature form.
- This recombinant protein can be used for biological assays. For research use only.