Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant R-spondin 11 (from HEK293 cells)
Human recombinant R-spondin 11 (from HEK293 cells)
Catalog # PRSI96-658
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant R-spondin 11 (from HEK293 cells)
Catalog # PRSI96-658
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilized human RSPO1 at 20μg/ml ( 100μl/well ) can bind human LIMPII with a linear ranger of 31. 25 - 1000 ng/mL.
  • Protein synonyms:
    CRISTIN3|R-spondin-1|RP11-566C13.1|FLJ40906|RSPO1
  • Protein/peptide name:
    R-Spondin 11
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    27.6 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

R-spondin-1 is also known as Roof plate-specific Spondin 1 (RSPO1) and cysteinerich and single thrombospondin domain containing protein 3 (Cristin 3), which is a secreted protein which belongs to the R-Spondin family and encodes a secreted activator protein with two cystein-rich, furin-like domains and one thrombospondin type 1 domain. All Rspondins regulate Wnt/β-catenin signaling, but have distinct expression patterns. Like other R-Spondins, R-Spondin-1 contains two adjacent cysteinerich furinlike domains (aa 34-135) with one potential N-glycosylation site, followed by a thrombospondin (TSP1) motif (aa 147-207) and a region rich in basic residues (aa 211-263). Only the furinlike domains are needed for β-catenin stabilisation. A putative nuclear localisation signal at the C-terminus may allow some expression in the nucleus.