Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant PD16 (from HEK293 cells)
Human recombinant PD16 (from HEK293 cells)
Catalog # PRSI96-611
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant PD16 (from HEK293 cells)
Catalog # PRSI96-611
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its binding ability in a functional ELISA against immobilized Mouse PD-L1, His Tag (Cat# PD1-M5220, 1 ug/well). The linear range is 0.16-5 ug/mL.
  • Protein synonyms:
    PD-1|hPD-1|hPD-l|CD279|PDCD1|PD1|SLEB2
  • Protein/peptide name:
    PD16
  • Purity:
    >92% as determined by SDS-PAGE.
  • Molecular weight:
    43.2 kDa
  • Formulation:
    Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
  • Tested applications:
    Western Blot

Specifications

About this item

Programmed cell death protein 1 (PD-1) is also known as CD279 and PDCD1, is a type I membrane protein and is a member of the extended CD28/CTLA-4 family of T cell regulators. PDCD1 is expressed on the surface of activated T cells, B cells, macrophages, myeloid cells and a subset of thymocytes. PD-1 has two ligands, PD-L1 and PD-L2, which are members of the B7 family. PD-L1 is expressed on almost all murine tumor cell lines, including PA1 myeloma, P815 mastocytoma, and B16 melanoma upon treatment with IFN-γ. PD-L2 expression is more restricted and is expressed mainly by DCs and a few tumor lines. PD1 inhibits the T-cell proliferation and production of related cytokines including IL-1, IL-4, IL-10 and IFN-γ by suppressing the activation and transduction of PI3K/AKT pathway. In addition, coligation of PD1 inhibits BCR-mediating signal by dephosphorylating key signal transducer. In vitro, treatment of anti-CD3 stimulated T cells with PD-L1-Ig results in reduced T cell proliferation and IFN-γ secretion. Monoclonal antibodies targeting PD-1 that boost the immune system are being developed for the treatment of cancer.