- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Protein synonyms:HM89|WHIM|NPYR|NPYRL|CXCR4|D2S201E|NPY3R|NPYY3R|LCR1|Fusin|HSY3RR|CD184|LAP3|LESTR|FB22
- Protein/peptide name:Cxcr4
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:32.8 kDa
- Formulation:Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
- Tested applications:Western Blot
C-X-C chemokine receptor type 4 is also known as fusin or CD184 (cluster of differentiation 184), CXCR4, CD184, D2S201E, FB22, HM89, HSY3RR, LAP3, LCR1, LESTR, NPY3R, NPYR, NPYRL, NPYY3R or WHIM. CXCR-4 is an alpha-chemokine receptor specific for stromal-derived-factor-1 (SDF-1 also called CXCL12), a molecule endowed with potent chemotactic activity for lymphocytes. This receptor is one of several chemokine receptors that HIV isolates can use to infect CD4+ T cells. HIV isolates that use CXCR4 are traditionally known as T-cell tropic isolates. Typically, these viruses are found late in infection. It is unclear as to whether the emergence of CXCR4 using HIV is a consequence or a cause of immunodeficiency.CXCR4 is upregulated during the implantation window in natural and hormone replacement therapy cycles in the endometrium, producing, in presence of a human blastocyst, a surface polarization of the CXCR4 receptors suggesting that this receptor is implicated in the adhesion phase of human implantation. SDF-1 and CXCR4 were believed to be a relatively "monogamous" ligand-receptor pair (other chemokines tend to use several different chemokine receptors in a fairly "promiscuous" manner). Recent evidence demonstrates ubiquitin is also a natural ligand of CXCR4. Chronic exposure to THC increased T lymphocyte CXCR4 expression on both CD4+ and CD8+ T lymphocytes. Drugs that block the CXCR4 receptor appear to be capable of "mobilizing" hematopoietic stem cells into the bloodstream as peripheral blood stem cells.