Bestelling invoeren
Netherlands
ContactUsLinkComponent
 
Human recombinant IL6 (fromE. coli)
Catalog # PRSI40-523
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL6 (fromE. coli)
Catalog # PRSI40-523
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • VPE:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Bron:
    E. coli
  • Species:
    Human
  • Biologische activiteiten:
    The ED50 was determined by the dose-dependent stimulation of the proliferation of the IL-6 dependent murine 7TD1 cells is ≤ 0.1 ng/mL, corresponding to a specific activity of ≥ 1 x 107 units/mg.
  • Protein synonyms:
    Interleukin-6|HSF|HGF|IFNB2|B-cell stimulatory factor 2|BSF2|IL-6
  • Protein/peptide name:
    IL6
  • Zuiverheid:
    >98%
  • Formulering:
    Lyophilized

Specifications

About this item

IL-6 is a pleiotropic cytokine that plays an important role in host defense by regulating immune and inflammatory responses. Produced by T cells, monocytes, fibroblasts, endothelial cells and keratinocytes, IL-6 has diverse biological functions. It stimulates B-cell differentiation and antibody production, synergizes with IL-3 in megakaryocyte development and platelet production, induces expression of hepatic acute-phase proteins, and regulates bone metabolism. IL-6 signals through the IL-6 receptor system that consists of two chains, IL-6R α and gp130. Murine IL-6 is inactive on human cells, while both human and murine are equally active on murine cells. Recombinant human IL-6 is a 20.9 kDa protein containing 184 amino acid residues. Recombinant rat IL-6 is a 21.7 kDa protein containing 187 amino acid residues. Recombinant murine IL-6 is a 21.7 kDa protein containing 187 amino acid residues.

  • Recombinant murine IL-6 is a 21.7 kDa protein containing 187 amino acid residues.