Human recombinant Pxf (from E. coli)
Catalog # PRSIXW-RP3215
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein synonyms:D1S2223E|PBD12A|PXMP1|OK/SW-cl.22|33 kDa housekeeping protein|PMPI|HK33|PXF|PMP1|Peroxisomal biogenesis factor 19
- Protein/peptide name:Pxf
- Purity:>95%
- Molecular weight:34.4 kDa (Calculated)
- Tested applications:ELISA, Western Blot, MS
Specifications
About this item
The covalent attachment of prenyl lipids, such as farnesyl or geranylgeranyl groups, by specific transferases is indispensable for the cellular sorting of many proteins. James et al. (1994) identified in hamster a farnesylated protein, called peroxisomal farnesylated protein or PxF, that localized to the outer surface of peroxisomes.
- This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.