Human recombinant Psat1 (from E. coli)
PRSIXW-RP3208
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:
- Protein synonyms:PSA|EPIP|Phosphohydroxythreonine aminotransferase|PSAT|Phosphoserine aminotransferase
- Protein/peptide name:Psat1
- Purity:>95%
- Molecular weight:42.6 kDa (Calculated)
- Tested applications:ELISA, Western Blot, MS
PSAT1 is likely a phosphoserine aminotransferase, based on similarity to proteins in mouse, rabbit, and Drosophila. It belongs to class-V of pyridoxal-phosphate-dependent aminotransferases and is required both in major phosphorylated pathway of serine biosynthesis and in the biosynthesis of pyridoxine.
- This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.