Order Entry
Netherlands
ContactUsLinkComponent
 
Human LXR-B peptide (14 aa near N-terminus)
  PRSI5579P
 :  
Human LXR-B peptide (14 aa near N-terminus)
  PRSI5579P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    14 aa near N-terminus
  • Protein/peptide name:
    LXR-B Peptide (14 aa near N-terminus)
  • Tested applications:
    Blocking

 

 

LXR-B peptide is used for blocking the activity of LXR-B antibody.