Order Entry
Netherlands
ContactUsLinkComponent
 
Human JMJD5 peptide (18 aa near C-terminus)
  PRSI5381P
 :  
Human JMJD5 peptide (18 aa near C-terminus)
  PRSI5381P
 :  

 

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    18 aa near C-terminus
  • Protein/peptide name:
    JMJD5 Peptide (18 aa near C-terminus)
  • Tested applications:
    Blocking

 

 

JMJD5 peptide is used for blocking the activity of JMJD5 antibody.