Human Hax1 isoform b peptide (10 aa near N-terminus)
PRSI4715P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:10 aa near N-terminus
- Protein/peptide name:Hax1 isoform b Peptide (10 aa near N-terminus)
- Tested applications:Blocking
Hax1b peptide is used for blocking the activity of Hax1b antibody.