Human Hax1 isoform a peptide (15 aa near N-terminus)
Catalog # PRSI4711P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near N-terminus
- Protein/peptide name:Hax1 isoform a Peptide (15 aa near N-terminus)
- Tested applications:Blocking
Specifications
About this item
Hax1a peptide is used for blocking the activity of Hax1a antibody.