Order Entry
Netherlands
ContactUsLinkComponent
Strep-tag II peptide
Strep-tag II peptide
Catalog # PRSI4335P
Supplier:  ProSci Inc.
CAS Number:  
undefined
Strep-tag II peptide
Catalog # PRSI4335P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Source:
  • Species:
  • Environmentally Preferable:
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Protein/peptide name:
    Strep-tag II Peptide
  • Tested applications:
    Blocking

Specifications

About this item

Strep-tag II peptide is used for blocking the activity of strep-tag II antibody.