Bestelling invoeren
Netherlands
ContactUsLinkComponent
HIV-1 gp120 HIV-1 gp120 Peptide (15 aa in the center)
HIV-1 gp120 HIV-1 gp120 Peptide (15 aa in the center)
Catalog # PRSI4777P
Supplier:  ProSci Inc.
CAS Number:  
undefined
HIV-1 gp120 HIV-1 gp120 Peptide (15 aa in the center)
Catalog # PRSI4777P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • VPE:
    50 µG
  • Species:
    HIV-1 gp120
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Aminozuurgetal:
    15 aa in the center
  • Protein/peptide name:
    HIV-1 gp120 Peptide (15 aa in the center)
  • Tested applications:
    Blocking

Specifications

About this item

gp120 peptide is used for blocking the activity of presenilin1 antibody.