Human CTRP6 peptide (15 aa near C-terminus)
PRSI3575P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near C-terminus
- Protein/peptide name:CTRP6 Peptide (15 aa near C-terminus)
- Tested applications:Blocking
CTRP6 peptide is used for blocking the activity of CTRP6 antibody.