Human caspase 8 isoform A peptide (16 aa near C-terminus)
Catalog # PRSI3473P
Leverancier: ProSci Inc.
CAS-nummer:
Specificaties
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:16 aa near C-terminus
- Protein/peptide name:Caspase 8 isoform A Peptide (16 aa near C-terminus)
- Tested applications:Blocking
Specificaties
Over dit artikel
Caspase-8 peptide is used for blocking the activity of Caspase-8 antibody.