Human BCL-xL peptide (14 aa near C-terminus)
PRSI3341P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa near C-terminus
- Protein/peptide name:BCL-xL Peptide (14 aa near C-terminus)
- Tested applications:Blocking
Bcl-xL peptide is used for blocking the activity of Bcl-xL antibody.