Human F1A alpha peptide (14 aa near C-terminus)
PRSI2279P
:
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa near C-terminus
- Protein/peptide name:F1A alpha Peptide (14 aa near C-terminus)
- Tested applications:Blocking
F1A alpha peptide is used for blocking the activity of F1A alpha antibody.