Anti-TNFRSF11A Rabbit Polyclonal Antibody (Biotin)
PRSIXP-5275BT
- Antigen name:Tumor Necrosis Factor Receptor Superfamily Member 11A
- Antigen symbol:TNFRSF11A
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:ODFR|RANK|OFE|OSTS|FEO|LOH18CR1|OPTB7|CD265|PDB2|TRANCER
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hsRANK Receptor (human sRANK Receptor).
- Size:50 µg
- Pk:50 µG
Anti-hsRANK Receptor specific antibody was purified by affinity chromatography and then biotinylated.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TNFRSF11A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human