Order Entry
Netherlands
ContactUsLinkComponent
Anti-GLUL Rabbit Polyclonal Antibody
Anti-GLUL Rabbit Polyclonal Antibody
Catalog # PRSI79-750
Supplier:  ProSci Inc.
undefined
Anti-GLUL Rabbit Polyclonal Antibody
Catalog # PRSI79-750
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Glutamate-Ammonia Ligase
  • Antigen symbol:
    GLUL
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    PIG43|GLNS|GS|PIG59
  • Concentration:
    1 mg/mL
  • Immunogen:
    Glutamine Synthetase antibody   was raised against a peptide sequence around aa.365~369 (G-D-E-P-F) derived from Human Glutamine Synthetase .
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

This enzyme has 2 functions: it catalyzes the production of glutamine and 4-aminobutanoate (gamma-aminobutyric acid, GABA), the latter in a pyridoxal phosphate-independent manner. Essential for proliferation of fetal skin fibroblasts.

Western Blot: 1:500~1:1000

Antibodies were purified by affinity-chromatography using epitope-specific peptide.

Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GLUL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat