Anti-PDPK1 Rabbit Polyclonal Antibody
Catalog # PRSI79-256
Supplier: ProSci Inc.
Specifications
- Antigen name:3-phosphoinositide dependent Protein kinase-1
- Antigen symbol:PDPK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:3 Phosphoinositide Dependent Protein Kinase 1|PkB kinase|PkB kinase like gene 1|PDPK1|PDK 1|hPDK 1|PDPK1_HUMAN.|PkB like 1
- Concentration:1 mg/mL
- Immunogen:PDK1 (Ab-241) antibody was raised against a peptide sequence around aa.239~243 (A-N-S-F-V) derived from Human PDK1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Phosphorylates and activates not only PKB/AKT, but also PKA, PKC-ζ, RPS6KA1 and RPS6KB1. May play a general role in signaling processes and in development
Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100, Immunofluorescence: 1:100~1:200
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PDPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat