Anti-HSP90AB1 Mouse Monoclonal Antibody
PRSI48-049
- Antigen name:Heat Shock Protein 90kDa Alpha (Cytosolic), Class B Member 1
- Antigen symbol:HSP90AB1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:HSP84|HSPCB|HSPC2|Heat shock protein HSP 90-beta|HSP 90|HSP90B|Heat shock 84 kDa|HSP 84|HSP90AB1
- Immunogen:Recombinant human TRA1 (676-803aa) purified from E. coli.
- Size:50 µg
- Pk:50 µG
Protein G Column
PBS, pH 7.4, 0.1% sodium azide
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: HSP90AB1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human