Anti-IL1R1 Mouse Monoclonal Antibody
Catalog # PRSI48-022
Supplier: ProSci Inc.
Specifications
- Antigen name:Interleukin 1 Receptor
- Antigen symbol:IL1R1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:P80|IL-1R-alpha|IL1R|D2S1473|CD121A|IL1RA
- Immunogen:The extracellular domain of the type I human IL-1 receptor.
- Tested applications:Radio Immunoassay
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Protein G Column
PBS, pH 7.4
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: IL1R1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human