Order Entry
Netherlands
ContactUsLinkComponent
Anti-IFNAR2 Goat Polyclonal Antibody
Anti-IFNAR2 Goat Polyclonal Antibody
Catalog # PRSI46-858
Supplier:  ProSci Inc.
undefined
Anti-IFNAR2 Goat Polyclonal Antibody
Catalog # PRSI46-858
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Interferon (Alpha, Beta And Omega) Receptor 2
  • Antigen symbol:
    IFNAR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human
  • Form:
    Liquid
  • Antigen synonyms:
    IFNARB|IFN-R|IFN-alpha-REC|IFNABR
  • Concentration:
    500 ug/mL
  • Immunogen:
    IFNAR2 antibody was raised against a 13 amino acid synthetic peptide near the internal region of IFNAR2.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

IFNAR2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

IFNAR2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IFNAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human