Anti-MARK4 Goat Polyclonal Antibody
Catalog # PRSI45-867
Supplier: ProSci Inc.
Specifications
- Antigen name:MAP/microtubule affinity-regulating kinase 4
- Antigen symbol:MARK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MAP/microtubule affinity-regulating kinase like 1|MARK4 serine/threonine protein kinase|MARK4|MAP/microtubule affinity-regulating kinase 4|MARKL1|KIAA1860|putative protein product of Nbla00650|Nbla00650|FLJ90097
- Concentration:500 ug/mL
- Immunogen:MARK4 antibody was raised against a 15 amino acid synthetic peptide near the C-Terminus of MARK4.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
MARK4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
MARK4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: MARK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human