Order Entry
Netherlands
ContactUsLinkComponent
Anti-EXO1 Goat Polyclonal Antibody
Anti-EXO1 Goat Polyclonal Antibody
Catalog # PRSI45-556
Supplier:  ProSci Inc.
undefined
Anti-EXO1 Goat Polyclonal Antibody
Catalog # PRSI45-556
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Exonuclease 1
  • Antigen symbol:
    EXO1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    HEX1|hExoI|EXOI|hExo1|EXO1|Exonuclease 1|Exonuclease I
  • Concentration:
    500 ug/mL
  • Immunogen:
    EXO1 antibody was raised against a 12 amino acid synthetic peptide near the internal region of EXO1.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

EXO1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

EXO1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EXO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human