Anti-EPB41L5 Goat Polyclonal Antibody
PRSI45-516
- Antigen name:Erythrocyte membrane Protein band 4.1 like L5
- Antigen symbol:EPB41L5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Form:Liquid
- Antigen synonyms:YMO1|YRT|BE37
- Concentration:500 ug/mL
- Immunogen:EBPL41L5 antibody was raised against a 13 amino acid synthetic peptide near the internal region of EBPL41L5.
- Size:100 µg
- Pk:100 µG
EBPL41L5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
EBPL41L5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EPB41L5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: