Anti-AOC3 Goat Polyclonal Antibody
Catalog # PRSI45-258
Supplier: ProSci Inc.
Specifications
- Antigen name:Amine Oxidase, Copper Containing 3
- Antigen symbol:AOC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:VAP-1.|Vascular adhesion protein 1|Semicarbazide-sensitive amine oxidase|Membrane primary amine oxidase|Membrane copper amine oxidase|Semicarbazide sensitive amine oxidase|Vascular Adhesion Protein 1|HPAO|SSAO|AOC3|VP97|AOC3_HUMAN|Amine Oxidase|Copper Containing 3 (vascular adhesion protein 1)|Copper amine oxidase
- Concentration:500 ug/mL
- Immunogen:AOC3 antibody was raised against a 13 amino acid synthetic peptide near the internal region (near the C-Terminus) of AOC3.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
AOC3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
AOC3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: AOC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human