Anti-CDH1 Goat Polyclonal Antibody
Catalog # PRSI42-929
Supplier: ProSci Inc.
Specifications
- Antigen name:Cadherin 1
- Antigen symbol:CDH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CD324|epithelial Cadherin|Cadherin-E|ECAD|Arc-1|CAM 120/80|cell-CAM 120/80|E-Cadherin|uvomorulin|Cadherin 1|Cadherin-1|E-Cadherin epithelial|calcium-dependent adhesion protein|epithelial|LCAM|CDHE|UVO
- Concentration:500 ug/mL
- Immunogen:CDH1 Antibody was raised against a 14 amino acid sequence near the internal region of CDH1 (aa662-675).
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
CDH1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CDH1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: