Anti-Pard3b Goat Polyclonal Antibody
PRSI42-922
- Antigen symbol:Pard3b
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Concentration:500 ug/mL
- Immunogen:Pard3b Antibody was raised against a 10 amino acid sequence near the C-Terminus of Pard3b (mouse).
- Size:100 µg
- Pk:100 µG
Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Pard3b antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Pard3b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse