Anti-GZMA Goat Polyclonal Antibody
PRSI42-290
- Antigen name:Granzyme A
- Antigen symbol:GZMA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Ganzyme 1 cytotoxic T-lymphocyte-associated serine esterase 3,Ctla-3|SE1|TSP-1|Hf|Ctla3|TSP1
- Concentration:500 ug/mL
- Immunogen:GZMA antibody was raised against an 11 amino acid synthetic peptide near the internal region of GZMA.
- Size:100 µg
- Pk:100 µG
GZMA antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
GZMA antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GZMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human