- Antigen name:Interleukin 6
- Antigen symbol:IL6
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:HSF|HGF|IFNB2|interferon beta 2|BSF2|IL-6
- Immunogen:Produced from sera of goats pre-immunized with highly pure (>98%) recombinant hIL-6. Human IL-6 specific antibody was purified by affinity chromatography and then biotinylated.
- Size:50 µg
- Pk:50 µG
Interleukin 6 (IL-6) is a multifunctional 24 kDa protein originally discovered in the medium of RNA stimulated fibroblastoid cells. It is upregulated by IL1, TNF, PDGF, IFN beta, TNF alpha, NGF, IL17 and downregulated by glucocorticoids IL4, TGF beta. IL-6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be as important as IL1 and TNF alpha in regulating the acute phase response. IL-6 has also been implicated in regulating adipose mass.
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type:
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human