Anti-ZBTB22 Rabbit Polyclonal Antibody
PRSI28-350
- Antigen name:Zinc Finger And BTB Domain Containing 22
- Antigen symbol:ZBTB22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:BING 1|ZBTB22A|Zinc finger and BTB domain containing protein 22|Zinc finger and BTB domain containing protein 22A|zinc finger and BTB domain containing 22|Fruitless|Protein BING1|Fru|ZNF297|Zinc finger protein 297|ZNF297A.
- Immunogen:A synthetic peptide corresponding a region of human ZBTB22.
- Size:50 µg
- Pk:50 µG
ZBTB22 contains 1 BTB (POZ) domain and 3 C2H2-type zinc fingers and belongs to the krueppel C2H2-type zinc-finger protein family. ZBTB22 may be involved in transcriptional regulation.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZBTB22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human