Anti-NET1 Rabbit Polyclonal Antibody
Catalog # PRSI26-632
Supplier: ProSci Inc.
Specifications
- Antigen name:Neuroepithelial Cell Transforming 1
- Antigen symbol:NET1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:ARHGEF8|NET1A|neuroepithelial cell transforming gene 1|NET1|Rho guanine nucleotide exchange factor (GEF) 8|p65 Net1 proto-oncogene protein|guanine nucleotide regulatory protein (oncogene)|small GTP-binding prot|neuroepithelioma transforming gene 1
- Immunogen:A synthetic peptide corresponding a region of human NET1.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
NET1 acts as guanine nucleotide exchange factor (GEF) for RhoA GTPase. It may be involved in activation of the SAPK/JNK pathway.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NET1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human