Specifications
- Antigen name:Coiled-coil-helix-coiled-coil-helix Domain Containing 4
- Antigen symbol:CHCHD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:TIMM40|MIA40
- Immunogen:A synthetic peptide corresponding a region of human CHCHD4.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
CHCHD4, a component of human mitochondria, belongs to a protein family whose members share 6 highly conserved cysteine residues constituting a -CXC-CX (9)C-CX (9)C- motif in the C terminus.CHCHD4, a component of human mitochondria, belongs to a protein family whose members share 6 highly conserved cysteine residues constituting a -CXC-CX (9)C-CX (9)C- motif in the C terminus (Hofmann et al., 2005 [PubMed 16185709]).
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CHCHD4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: