Anti-FBXO7 Rabbit Polyclonal Antibody
Product Details & Documents
FBXO7 is a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. FBXO7 belongs to the Fbxs class and it may play a role in regulation of hematopoiesis.This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it may play a role in regulation of hematopoiesis. Alternatively spliced transcript variants of this gene have been identified with the full-length natures of only some variants being determined.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FBXO7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
10102-688.pdf
Datasheet
Specifications
- Size:50 µg
- Western blot:Yes
- Cat. No.:PRSI25-825
- Clonality:Polyclonal
- Reactivity:Human
- Form:Lyophilized
- Antigen synonyms:PKPS|F box protein 7|F-box protein 7|FBX07|FBX7|PARK15|F box only protein 7|FBX
- Immunogen:A synthetic peptide corresponding a region of human FBXO7.
- Host:Rabbit
- Conjugation:Unconjugated
- Antigen symbol:FBXO7
- Antigen name:F-box Protein 7
- ELISA:Yes



